1,2,3,4-Butanetetracarboxylic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.14924.0050 8.14924.0251 8.14924.0050 8.14924.0251
EM8.14924.0050CS 7279.06 CAD
EM8.14924.0050 EM8.14924.0251 CA8.14924.0050 CA8.14924.0251
1,2,3,4-Butanetetracarboxylic acid for synthesis, Sigma-Aldrich®
1,2,3,4-Butanetetracarboxylic acid
Formula: C₈H₁₀O₈
MW: 234.16 g/mol
MDL Number: MFCD00002722
CAS Number: 1703-58-8

Order Now

Specification Test Results

Solubility 118 g/l (19 °C)
Melting Point 187 - 190 °C
Molar Mass 234.16 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR