2-Methyl-2-butene mixture of isomers 83,0-88,0 % (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Trimethylethylene, Isoamylene, Isopentene

M-Clarity™ quality level = MQ200

8.14874.0025 8.14874.0250 8.14874.1000 8.14874.9025 8.14874.0025 8.14874.0250 8.14874.1000 8.14874.9025
EM8.14874.0025CS 8996.52 CAD
EM8.14874.0025 EM8.14874.0250 EM8.14874.1000 EM8.14874.9025 CA8.14874.0025 CA8.14874.0250 CA8.14874.1000 CA8.14874.9025
2-Methyl-2-butene mixture of isomers 83,0-88,0 % (by GC) for synthesis, Sigma-Aldrich®
2-Methyl-2-butene
Formula: C₅H₁₀
MW: 70.13 g/mol
Melting Pt: –134 °C
Density: 0.66 g/cm3 (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009276
CAS Number: 513-35-9
UN: 3295
ADR: 3,I

Order Now

Specification Test Results

Solubility 0.19 g/l (25 °C)
Melting Point -134 °C
Molar Mass 70.13 g/mol
Vapor Pressure 513 hPa (20 °C)
Flash Point -45 °C
Explosion Limit 1 - 9 %(V)
Density 0.66 g/cm3 (20 °C)
Ignition Point 290 °C
Viscosity Kinematic 0.29 mm2/s (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR