Sodium chlorite 25% in water for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.14815.0250 8.14815.0250 8.14815.1000
EM8.14815.0250CS 782.1 CAD
EM8.14815.0250 CA8.14815.0250 CA8.14815.1000
Sodium chlorite 25% in water for synthesis, Sigma-Aldrich®
Sodium chlorite
Formula: ClNaO₂
MW: 90.44 g/mol
MDL Number: MFCD00003478
CAS Number: 7758-19-2
UN: 1908
ADR: 8,II

Order Now

Specification Test Results

Boiling Point 100 °C
Density 1.21 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR