Isopropyl 2-bromo-2-methylpropionate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Isopropyl 2-bromo-2-methylpropanoate, Isopropyl 2-bromoisobutyrate, 2-Bromo-2-methylpropionic Acid Isopropyl Ester

M-Clarity™ quality level = MQ200

8.14785.0025 8.14785.0100 8.14785.0025 8.14785.0100
EM8.14785.0025CS 12822.65 CAD
EM8.14785.0025 EM8.14785.0100 CA8.14785.0025 CA8.14785.0100
Isopropyl 2-bromo-2-methylpropionate for synthesis, Sigma-Aldrich®
Isopropyl 2-bromo-2-methylpropionate
Formula: C₇H₁₃BrO₂
MW: 209.08 g/mol
MDL Number: MFCD00055235
CAS Number: 51368-55-9

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 209.08 g/mol
Boiling Point 170 °C
Flash Point 63 °C
Density 1.24 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR