n-Tetradecane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Tetradecane

M-Clarity™ quality level = MQ200

8.14767.0050 8.14767.0250 8.14767.0050 8.14767.0250
EM8.14767.0050CS 4795.3 CAD
EM8.14767.0050 EM8.14767.0250 CA8.14767.0050 CA8.14767.0250
n-Tetradecane for synthesis, Sigma-Aldrich®
n-Tetradecane
Formula: H₃C(CH₂)₁₂CH₃
MW: 198.39 g/mol
MDL Number: MFCD00008986
CAS Number: 629-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03064a29015d1e810f1e4350a7a6cd2dfe4713fd&vid=f4838a5686fe033622495a3bf280963555654d9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility  - 0.0001 g/l (20 °C)
Melting Point 6 °C
Molar Mass 198.39 g/mol
Boiling Point 254 °C
Vapor Pressure 1.55 Pa (25 °C)
Flash Point 102 °C
Explosion Limit 0.5 - 6.5 %(V)
Density 0.76 g/cm3 (20 °C)
Ignition Point 202 °C
Viscosity Kinematic 3 mm2/s (40 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR