Terpineol, mixture of isomers for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.14759.1000 8.14759.2500 8.14759.1000 8.14759.2500
EM8.14759.1000CS 1613.18 CAD
EM8.14759.1000 EM8.14759.2500 CA8.14759.1000 CA8.14759.2500
Terpineol, mixture of isomers for synthesis, Sigma-Aldrich®
Terpineol, mixture of isomers
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00166983
CAS Number: 8000-41-7

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 154.25 g/mol
Boiling Point 214 - 217 °C
Vapor Pressure 0.24 hPa (20 °C)
Flash Point 91 °C
Density 0.930 - 0.935 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR