Octamethylcyclotetrasiloxane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.14750.0025 8.14750.0100 8.14750.0250 8.14750.0100 8.14750.0250
EM8.14750.0025CS 5825.46 CAD
EM8.14750.0025 EM8.14750.0100 EM8.14750.0250 CA8.14750.0100 CA8.14750.0250
Octamethylcyclotetrasiloxane for synthesis, Sigma-Aldrich®
Octamethylcyclotetrasiloxane
Formula: [-Si(CH₃)₂O-]₄
MDL Number: MFCD00003269
CAS Number: 556-67-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Solubility  - 0.001 g/l (25 °C)
Melting Point 18 °C
Molar Mass 296.62 g/mol
Boiling Point 176 °C
Vapor Pressure 1.3 hPa (20 °C)
Flash Point 51 °C
Explosion Limit 0.4 - 11.7 %(V)
Density 0.95 g/cm3
Ignition Point 400 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR