Diisobutylene mixed isomers zS, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2,4,4-Trimethylpentene, Isooctene

8.14734.0100 8.14734.1000 8.14734.0100 8.14734.1000
EM8.14734.0100CS 1038.06 CAD
EM8.14734.0100 EM8.14734.1000 CA8.14734.0100 CA8.14734.1000
Diisobutylene mixed isomers zS, Sigma-Aldrich®
Diisobutylene
Formula: C₈H₁₆
MW: 112.22 g/mol
Melting Pt: -106
Flash Pt: -6
MDL Number: MFCD00008855
CAS Number: 25167-70-8
UN: 2050
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf11704306b612da2bff59a5038c5c1c0b8275f9&vid=3747a312c11312949e0c54a3dd52e9052e14ca16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility  - 0.1 g/l (20 °C)
Melting Point -106 °C
Molar Mass 112.21 g/mol
Boiling Point 98 - 105 °C (1013 hPa)
Vapor Pressure 41 hPa (20 °C)
Flash Point -12 °C
Refractive Index 1.411
Explosion Limit 0.7 - 5.8 %(V)
Density 0.72 g/cm3 (20 °C)
pH 7 (H2O)
Ignition Point 380 °C DIN 51794
Viscosity Kinematic 0.76 mm2/s (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR