Dimethoxy(diisobutyl)silane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
|
Synonyms:
Diisobutyldimethoxysilane
M-Clarity™ quality level = MQ200
8.14720.0025
8.14720.0100
8.14720.0025
8.14720.0100
EM8.14720.0025CS
9988.76
CAD
EM8.14720.0025
EM8.14720.0100
CA8.14720.0025
CA8.14720.0100
Dimethoxy(diisobutyl)silane for synthesis, Sigma-Aldrich®
Dimethoxy(diisobutyl)silane
Formula:
C₁₀H₂₄O₂Si MW: 204.38 g/mol |
MDL Number:
MFCD00236510 CAS Number: 17980-32-4 UN: 3082 ADR: 9,III |
Specification Test Results
Solubility | 13.4 mg/l (25 °C) Hydrolysis |
Molar Mass | 204.38 g/mol |
Boiling Point | 188 °C (1025 hPa) |
Vapor Pressure | 0.75 hPa (25 °C) |
Flash Point | 62 °C |
Density | 0.863 g/cm3 (20 °C) |
Ignition Point | 255 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...