Trifluoroacetamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2,2,2-Trifluoroacetamide

M-Clarity™ quality level = MQ200

8.14690.0010 8.14690.0050 8.14690.0010 8.14690.0050
EM8.14690.0010CS 32141.56 CAD
EM8.14690.0010 EM8.14690.0050 CA8.14690.0010 CA8.14690.0050
Trifluoroacetamide for synthesis, Sigma-Aldrich®
Trifluoroacetamide
Formula: C₂H₂F₃NO
MW: 113.04 g/mol
Boiling Pt: 162.4
MDL Number: MFCD00008008
CAS Number: 354-38-1

Order Now

Specification Test Results

Solubility 460 g/l (23 °C)
Melting Point 73 - 75 °C
Molar Mass 113.03 g/mol
Bulk Density 960 kg/m3
Boiling Point 162.5 °C
pH 3.6 (460 g/l, H2O, 23 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR