Formamidine acetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.14653.0100 8.14653.0500 8.14653.0100 8.14653.0500
EM8.14653.0100CS 14333.76 CAD
EM8.14653.0100 EM8.14653.0500 CA8.14653.0100 CA8.14653.0500
Formamidine acetate for synthesis, Sigma-Aldrich®
Formamidine acetate
Formula: C₃H₈N₂O₂
MDL Number: MFCD00012866
CAS Number: 3473-63-0

Order Now

Specification Test Results

Solubility 832 g/l (20 °C)
Melting Point 156 - 160 °C
Molar Mass 104.11 g/mol
pH 8 (400 g/l, H2O, 20 °C)
Ignition Point 565 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR