Betaine hydrochloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.14633.0100 8.14633.1000 8.14633.0100 8.14633.1000
EM8.14633.0100CS 7407.04 CAD
EM8.14633.0100 EM8.14633.1000 CA8.14633.0100 CA8.14633.1000
Betaine hydrochloride for synthesis, Sigma-Aldrich®
Betaine hydrochloride
Formula: C₅H₁₂ClNO₂
MDL Number: MFCD00011903
CAS Number: 590-46-5

Order Now

Specification Test Results

Solubility 647 g/l (25 °C)
Melting Point 242 - 246 °C
Molar Mass 153.61 g/mol
pH 1 (50 g/l, H2O, 20 °C)
Ignition Point 400 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR