L(+)-Glutamic acid hydrochloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.14632.0250 8.14632.9025 8.14632.0250 8.14632.9025
EM8.14632.0250CS 1191.32 CAD
EM8.14632.0250 EM8.14632.9025 CA8.14632.0250 CA8.14632.9025
L(+)-Glutamic acid hydrochloride for synthesis, Sigma-Aldrich®
L(+)-Glutamic acid hydrochloride
Formula: C₅H₁₀ClNO₄
Melting Pt: 191…193 °C (dec.)
Density: 1.525 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00012619
CAS Number: 138-15-8

Order Now

Specification Test Results

Solubility 490 g/l (20 °C)
Melting Point 191 - 193 °C decomposes
Molar Mass 183.59 g/mol
Bulk Density 630 kg/m3
Density 1.525 g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR