2-(Trimethylsiloxy)-1,3-butadiene for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.14551.0010
8.14551.0050
8.14551.0010
8.14551.0050
EM8.14551.0010CS
129962.9
CAD
EM8.14551.0010
EM8.14551.0050
CA8.14551.0010
CA8.14551.0050
2-(Trimethylsiloxy)-1,3-butadiene for synthesis, Sigma-Aldrich®
2-(Trimethylsiloxy)-1,3-butadiene
Formula:
C₇H₁₄OSi |
MDL Number:
MFCD00009852 CAS Number: 38053-91-7 UN: 1993 ADR: 3,II |
Specification Test Results
Solubility | (20 °C) insoluble |
Molar Mass | 142.27 g/mol |
Boiling Point | 124 °C (1013 hPa) |
Flash Point | 12 °C |
Density | 0.82 g/cm3 (20 °C) |
Ignition Point | 350 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...