Formaldehyde diethyl acetal stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Ethylal, Diethoxymethane

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
M-Clarity™ quality level = MQ200

8.14549.0250 8.14549.1000 8.14549.0250 8.14549.1000
EM8.14549.0250CS 759.98 CAD
EM8.14549.0250 EM8.14549.1000 CA8.14549.0250 CA8.14549.1000
Formaldehyde diethyl acetal stabilized for synthesis, Sigma-Aldrich®
Formaldehyde diethyl acetal
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 88
Melting Pt: -66
Flash Pt: -6
MDL Number: MFCD00009246
CAS Number: 462-95-3
UN: 2373
ADR: 3 II

Order Now

Specification Test Results

Solubility 91 g/l (18 °C)
Melting Point -66 °C
Molar Mass 104.15 g/mol
Boiling Point 88 °C (1013 hPa)
Vapor Pressure 70 hPa (20 °C)
Flash Point -6 °C
Explosion Limit 1.5 %(V)
Density 0.83 g/cm3 (20 °C)
Ignition Point 174 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR