Chlorobenzene, special quality for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene

M-Clarity™ quality level = MQ200

8.14506.1000 8.14506.2500 8.14506.1000 8.14506.2500
EM8.14506.1000CS 993.82 CAD
EM8.14506.1000 EM8.14506.2500 CA8.14506.1000 CA8.14506.2500
Chlorobenzene, special quality for synthesis, Sigma-Aldrich®
Chlorobenzene
Formula: C₆H₅Cl
MDL Number: MFCD00000530
CAS Number: 108-90-7
UN: 1134
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.4 g/l (20 °C)
Melting Point -45 °C
Molar Mass 112.56 g/mol
Boiling Point 132 °C (1013 hPa)
Vapor Pressure 12 hPa (20 °C)
Flash Point 27 °C
Refractive Index 1.5248 (20 °C, 589 nm)
Explosion Limit 1.3 - 11 %(V)
Density 1.11 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 590 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR