3-Amino-1H-1,2,4-triazole for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.14495.0100 8.14495.0500 8.14495.0100 8.14495.0500
EM8.14495.0100CS 14243.7 CAD
EM8.14495.0100 EM8.14495.0500 CA8.14495.0100 CA8.14495.0500
3-Amino-1H-1,2,4-triazole for synthesis, Sigma-Aldrich®
3-Amino-1H-1,2,4-triazole
Formula: C₂H₄N₄
MDL Number: MFCD00053362
CAS Number: 61-82-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility 280 g/l (25 °C)
Melting Point 149 - 152 °C
Molar Mass 84.08 g/mol
Bulk Density 650 kg/m3
Vapor Pressure  - 0.1 hPa (20 °C)
Density 1.138 g/cm3 (20 °C)
pH 6.4 - 7.4 (100 g/l, H2O, 20 °C)
Ignition Point 225 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR