Samarium, powder for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.14479.0001 8.14479.0010 8.14479.0001 8.14479.0010
EM8.14479.0001CS 6447.98 CAD
EM8.14479.0001 EM8.14479.0010 CA8.14479.0001 CA8.14479.0010
Samarium, powder for synthesis, Sigma-Aldrich®
Samarium
Formula: Sm
MW: 150.36 g/mol
MDL Number: MFCD00011233
CAS Number: 7440-19-9
UN: 3208
ADR: 4.3,I

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 1072 °C
Molar Mass 150.36 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR