Gibberellic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid
M-Clarity™ quality level = MQ200
8.14464.0001
8.14464.0005
8.14464.8500
8.14464.0001
8.14464.8500
EM8.14464.0001CS
8822.72
CAD
EM8.14464.0001
EM8.14464.0005
EM8.14464.8500
CA8.14464.0001
CA8.14464.0005
CA8.14464.8500
Gibberellic acid for synthesis, Sigma-Aldrich®
Gibberellic acid
Formula:
C₁₉H₂₂O₆ MW: 346.38 g/mol |
MDL Number:
MFCD00079329 CAS Number: 77-06-5 |
Specification Test Results
Solubility | 5 g/l (25 °C) |
Melting Point | 233 - 235 °C |
Molar Mass | 346.37 g/mol |
Density | 1.34 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...