1-Octadecanethiol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
n-Octadecyl mercaptan, n-Octadecanethiol, Stearyl mercaptane, 1-Mercaptooctadecane, Octadecane-1-thiol
M-Clarity™ quality level = MQ200
8.14444.0050
8.14444.8000
8.14444.0050
8.14444.8000
EM8.14444.0050CS
9393.1
CAD
EM8.14444.0050
EM8.14444.8000
CA8.14444.0050
CA8.14444.8000
1-Octadecanethiol for synthesis, Sigma-Aldrich®
1-Octadecanethiol
Formula:
C₁₈H₃₈S MW: 286.57 g/mol Boiling Pt: 204 |
MDL Number:
MFCD00004886 CAS Number: 2885-00-9 UN: 3335 ADR: 9 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 28 - 31 °C |
Molar Mass | 286.55 g/mol |
Boiling Point | 366 °C |
Vapor Pressure |
|
Flash Point | 185 °C |
Refractive Index | 1.4645 (20 °C, 589 nm) |
Density | 0.847 g/cm3 (20 °C) |
Ignition Point |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...