Potassium hydroxide, powder for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.14353.1000 8.14353.0100 8.14353.1000 8.14353.5000 8.14353.9050 8.14353.0100 8.14353.5000 8.14353.9050
CA8.14353.1000EA 94 CAD
CA8.14353.1000 EM8.14353.0100 EM8.14353.1000 EM8.14353.5000 EM8.14353.9050 CA8.14353.0100 CA8.14353.5000 CA8.14353.9050
Potassium hydroxide, powder for synthesis, Sigma-Aldrich®
Potassium hydroxide
Formula: KOH
Melting Pt: 361 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8,II

Order Now

Specification Test Results

Solubility 1130 g/l (20 °C)
Melting Point 360 °C
Molar Mass 56.11 g/mol
Boiling Point 1320 °C
Vapor Pressure (20 °C)
Density 2.04 g/cm3 (20 °C)
pH 14 (56 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR