Silver trifluoromethanesulfonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Silver trifluoromethanesulphonate, Silver triflate, Silver trifluoromethanesulfonate, Trifluoromethanesulphonic acid silver salt, Trifluoromethanesulfonic acid silver salt

M-Clarity™ quality level = MQ200

8.14325.0005 8.14325.0025 8.14325.0005 8.14325.0025
EM8.14325.0005CS 150430.22 CAD
EM8.14325.0005 EM8.14325.0025 CA8.14325.0005 CA8.14325.0025
Silver trifluoromethanesulfonate for synthesis, Sigma-Aldrich®
Silver trifluoromethanesulphonate
Formula: CAgF₃O₃S
MDL Number: MFCD00013226
CAS Number: 2923-28-6
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 356 °C
Molar Mass 256.94 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR