Bromotrimethylsilane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Trimethylbromosilane, Trimethylsilyl bromide
M-Clarity™ quality level = MQ200
8.14324.0010
8.14324.0100
8.14324.0010
8.14324.0100
EM8.14324.0010CS
122363.1
CAD
EM8.14324.0010
EM8.14324.0100
CA8.14324.0010
CA8.14324.0100
Bromotrimethylsilane for synthesis, Sigma-Aldrich®
Bromotrimethylsilane
Formula:
C₃H₉BrSi MW: 153.09 g/mol Boiling Pt: 79 Melting Pt: -43 |
MDL Number:
MFCD00000048 CAS Number: 2857-97-8 UN: 2920 ADR: 8 (3) II |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Melting Point | -43 °C |
Molar Mass | 153.09 g/mol |
Boiling Point | 79 °C |
Flash Point | 32 °C |
Refractive Index | 1.4211 (25 °C, 589 nm) |
Density | 1.17 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...