Triphosgene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Bis(trichloromethyl) carbonate

M-Clarity™ quality level = MQ200

8.14283.0025 8.14283.0100 8.14283.1000 8.14283.0025 8.14283.0100 8.14283.1000
EM8.14283.0025CS 18745.12 CAD
EM8.14283.0025 EM8.14283.0100 EM8.14283.1000 CA8.14283.0025 CA8.14283.0100 CA8.14283.1000
Triphosgene for synthesis, Sigma-Aldrich®
Triphosgene
Formula: C₃Cl₆O₃
MW: 296.75 g/mol
MDL Number: MFCD00062848
CAS Number: 32315-10-9
UN: 2811
ADR: 6.1,I

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 77 - 81 °C
Molar Mass 296.75 g/mol
Boiling Point 105 °C (30 hPa)
Vapor Pressure 16 hPa (90 °C)
Density 1.6 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR