Levulinic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 4-Oxovaleric acid, 4-Ketovaleric acid, 4-Oxopentanoic acid

M-Clarity™ quality level = MQ200

8.14254.0100 8.14254.0500 8.14254.0100 8.14254.0500
EM8.14254.0100CS 9513.18 CAD
EM8.14254.0100 EM8.14254.0500 CA8.14254.0100 CA8.14254.0500
Levulinic acid for synthesis, Sigma-Aldrich®
Levulinic acid
Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 245
Flash Pt: 138
MDL Number: MFCD00002796
CAS Number: 123-76-2
UN: 3261
Merck Index: 13,05492

Order Now

Specification Test Results

Solubility 675 g/l (25 °C)
Melting Point 26 - 29 °C
Molar Mass 116.12 g/mol
Boiling Point 245 - 246 °C (1013 mbar)
Flash Point 98 °C
Refractive Index 1.4396 (20 °C, 589 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR