3-Diethylaminopropylamine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
3-Diethylamino-1-propylamine, DEAPA, N,N-Diethyl-1,3-propanediamine, N,N-Diethyl-1,3-diaminopropane, 3-Aminopropyldiethylamine, 3-(Diethylamino)propylamine
M-Clarity™ quality level = MQ200
8.14100.0100
8.14100.1000
8.14100.0100
8.14100.1000
EM8.14100.0100CS
959.06
CAD
EM8.14100.0100
EM8.14100.1000
CA8.14100.0100
CA8.14100.1000
3-Diethylaminopropylamine for synthesis, Sigma-Aldrich®
3-Diethylaminopropylamine
Formula:
C₇H₁₈N₂ MW: 130.23 g/mol Melting Pt: -60 Flash Pt: 51 |
MDL Number:
MFCD00008218 CAS Number: 104-78-9 UN: 2684 ADR: 3 (8) III |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -60 °C |
Molar Mass | 130.23 g/mol |
Boiling Point | 159.0 °C |
Vapor Pressure | 2.2 hPa (20 °C) |
Flash Point | 51 °C |
Refractive Index | 1.4413 (20 °C, 589 nm) |
Explosion Limit | 1.0 %(V) |
Density | 0.82 g/cm3 (20 °C) |
pH | 11.7 (10 g/l, H2O, 20 °C) |
Ignition Point | 215 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...