Zirconium tetrachloride, anhydrous for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Zirconium (IV) chloride

M-Clarity™ quality level = MQ200

8.08913.0005 8.08913.0250 8.08913.1000 8.08913.5000 8.08913.0005 8.08913.0250 8.08913.1000 8.08913.5000
EM8.08913.0005CS 7045.22 CAD
EM8.08913.0005 EM8.08913.0250 EM8.08913.1000 EM8.08913.5000 CA8.08913.0005 CA8.08913.0250 CA8.08913.1000 CA8.08913.5000
Zirconium tetrachloride, anhydrous for synthesis, Sigma-Aldrich®
Zirconium tetrachloride
Formula: Cl₄Zr
MDL Number: MFCD00011306
CAS Number: 10026-11-6
UN: 2503
ADR: 8,III

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 437 °C
Molar Mass 233.04 g/mol
Bulk Density 1200 kg/m3
Vapor Pressure 1.3 hPa (190 °C)
Density 2.80 g/cm3 (15 °C)
pH  - 1 (H2O, 20 °C) (saturated aqueous solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR