o-Xylene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1,2-Dimethylbenzene

M-Clarity™ quality level = MQ200

8.08697.1000 8.08697.6010 8.08697.6010 8.08697.2500 8.08697.1000 8.08697.2500
EM8.08697.1000CS 1266.12 CAD
EM8.08697.1000 CA8.08697.2500 EM8.08697.6010 CA8.08697.1000 EM8.08697.2500 CA8.08697.6010
o-Xylene for synthesis, Sigma-Aldrich®
o-Xylene
Formula: C₈H₁₀
MW: 106.17 g/mol
MDL Number: MFCD00008519
CAS Number: 95-47-6
UN: 1307
ADR: 3,III
https://ca.vwr.com/stibo/web/std.lang.all/85/44/768544.jpg

Order Now

Specification Test Results

Solubility 0.18 g/l (20 °C)
Melting Point -25 °C
Molar Mass 106.17 g/mol
Boiling Point 144.4 °C (1013 hPa)
Vapor Pressure 7 hPa (20 °C)
Flash Point 30 °C
Explosion Limit 1.0 - 7.6 %(V)
Density 0.88 g/cm3 (20 °C)
Ignition Point 465 °C
Evaporation Number 13.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR