p-Xylene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1,4-Dimethylbenzene

M-Clarity™ quality level = MQ200

8.08691.1000 8.08691.2500 8.08691.1000 8.08691.2500
EM8.08691.1000CS 756.82 CAD
EM8.08691.1000 EM8.08691.2500 CA8.08691.1000 CA8.08691.2500
p-Xylene for synthesis, Sigma-Aldrich®
p-Xylene
Formula: C₆H₄(CH₃)₂
MDL Number: MFCD00008556
CAS Number: 106-42-3
UN: 1307
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8b04563a5941cefd103ebd3048bcf8486f2bdac&vid=e5db83ad1f0cb6a583610c58cea291dcbe1be1f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.2 g/l (25 °C)
Melting Point 13.3 °C
Molar Mass 106.17 g/mol
Boiling Point 138.4 °C (1013 hPa)
Vapor Pressure 8.7 hPa (20 °C)
Flash Point 24 °C
Explosion Limit 1.1 - 7 %(V)
Density 0.86 g/cm3 (20 °C)
Ignition Point 540 °C
Evaporation Number 13.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR