1,3-Diaminopropane ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.08272.0100 8.08272.0500 8.08272.7500 8.08272.9045 8.08272.0100 8.08272.0500 8.08272.7500 8.08272.9045
EM8.08272.0100CS 1038.06 CAD
EM8.08272.0100 EM8.08272.0500 EM8.08272.7500 EM8.08272.9045 CA8.08272.0100 CA8.08272.0500 CA8.08272.7500 CA8.08272.9045
1,3-Diaminopropane ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
1,3-Diaminopropane
Formula: C₃H₁₀N₂
Boiling Pt: 137…140 °C (1013 mbar)
Melting Pt: –12 °C
Density: 0.89 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008228
CAS Number: 109-76-2
UN: 2922
ADR: 8,6.1,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -12 °C
Molar Mass 74.12 g/mol
Boiling Point 137 - 140 °C (1013 mbar)
Vapor Pressure  - 10 hPa (20 °C)
Flash Point 50 °C
Refractive Index 1.4575 (20 °C, 589 nm)
Explosion Limit 2.8 - 15.2 %(V)
Density 0.89 g/cm3 (20 °C)
pH 12 (100 g/l, H2O, 20 °C)
Ignition Point 350 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR