Toluenediisocyanate mixture of isomers ≥98% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.08264.0100 8.08264.0500 8.08264.2500 8.08264.9025 8.08264.0100 8.08264.0500 8.08264.2500 8.08264.9025
EM8.08264.0100CS 1022.26 CAD
EM8.08264.0100 EM8.08264.0500 EM8.08264.2500 EM8.08264.9025 CA8.08264.0100 CA8.08264.0500 CA8.08264.2500 CA8.08264.9025
Toluenediisocyanate mixture of isomers ≥98% for synthesis, Sigma-Aldrich®
Toluenediisocyanate

Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Formula: C₉H₆N₂O₂
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 12…14 °C
Density: 1.22 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01778251
CAS Number: 26471-62-5
EINECS: 247-722-4
UN: 2078
ADR: 6.1,II

Order Now

Specification Test Results

Assay isomers (GC, area%) ≥ 98.0 % (a/a)
Assay Toluylene-2,6-diisocyanate (NMR) 10.0 - 30.0 % (a/a)
Assay Toluylene-2,4-diisocyanate (NMR) 70.0 - 90.0 % (a/a)
Density (d 20 °C/ 4 °C) 1.218 - 1.222
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR