Methyltrichlorosilane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Trichloro(methyl)silane

M-Clarity™ quality level = MQ200

8.08255.0250 8.08255.1000 8.08255.0250 8.08255.1000
EM8.08255.0250CS 729.96 CAD
EM8.08255.0250 EM8.08255.1000 CA8.08255.0250 CA8.08255.1000
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Methyltrichlorosilane
Formula: CH₃Cl₃Si
MW: 149.48 g/mol
MDL Number: MFCD00000481
CAS Number: 75-79-6
UN: 1250
ADR: 3 (8),II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point -90 °C
Molar Mass 149.47 g/mol
Boiling Point 66 °C
Vapor Pressure 189 hPa (20 °C)
Flash Point -1 °C
Refractive Index 1.411 (20 °C, 589 nm)
Explosion Limit 5.5 - 10.4 %(V)
Density 1.27 g/cm3 (20 °C)
pH (H2O) acidic,Hydrolysis
Ignition Point 500 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR