Triisopropanolamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 1,1',1''-Nitrilotripropan-2-ol, 1,1',1"-Nitrilotri-2-propanol, Tris(2-hydroxypropyl)amine

M-Clarity™ quality level = MQ200

8.08236.0100 8.08236.1000 8.08236.0100 8.08236.1000
EM8.08236.0100CS 2493.24 CAD
EM8.08236.0100 EM8.08236.1000 CA8.08236.0100 CA8.08236.1000
Triisopropanolamine for synthesis, Sigma-Aldrich®
Triisopropanolamine
Formula: C₉H₂₁NO₃
MW: 191.27 g/mol
MDL Number: MFCD00004533
CAS Number: 122-20-3

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 50 °C
Molar Mass 191.27 g/mol
Boiling Point 305 °C (1013 hPa)
Vapor Pressure 1 hPa (100 °C)
Flash Point 160 °C
Explosion Limit 0.8 - 5.8 %(V)
Density 1.01 g/cm3 (50 °C)
pH 10.8 (100 g/l, H2O, 20 °C)
Ignition Point 275 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR