Toluene-4-sulfonamide ≥98.0% (by HPLC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: p-Toluenesulfonamide, 4-Methylbenzenesulfonamide, PTSA, 4-Toluenesulfonamide, p-Toluenesulphonamide, 4-Methylbenzenesulphonamide, 4-Toluenesulphonamide

M-Clarity™ quality level = MQ200

8.08230.0100 8.08230.1000 8.08230.9025 8.08230.0100 8.08230.1000 8.08230.9025
EM8.08230.0100CS 677.82 CAD
EM8.08230.0100 EM8.08230.1000 EM8.08230.9025 CA8.08230.0100 CA8.08230.1000 CA8.08230.9025
Toluene-4-sulfonamide ≥98.0% (by HPLC) for synthesis, Sigma-Aldrich®
Toluene-4-sulphonamide
Formula: C₇H₉NO₂S
MW: 171.22 g/mol
Boiling Pt: 220…222 °C (13 hPa)
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00011692
CAS Number: 70-55-3

Order Now

Specification Test Results

Solubility 3200 mg/l (25 °C)
Melting Point 136 - 138 °C
Molar Mass 171.21 g/mol
Boiling Point 220 - 222 °C (13 hPa)
Flash Point 202 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR