Thioacetic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.08076.0100 8.08076.0250 8.08076.0100 8.08076.0250
EM8.08076.0100CS 1738 CAD
EM8.08076.0100 EM8.08076.0250 CA8.08076.0100 CA8.08076.0250
Thioacetic acid for synthesis, Sigma-Aldrich®
Thioacetic acid
Formula: C₂H₄OS
MDL Number: MFCD00004853
CAS Number: 507-09-5
UN: 2436
ADR: 3,II

Order Now

Specification Test Results

Solubility 27 g/l (15 °C) (slow decomposition)
Melting Point -17 °C
Molar Mass 76.12 g/mol
Boiling Point 88 - 92 °C (1013 hPa)
Vapor Pressure 1 hPa (20 °C)
Flash Point 18 °C
Refractive Index 1.463 (20 °C, 589 nm)
Density 1.07 g/cm3 (20 °C)
pH 1.8 (100 g/l, H2O, 20 °C) Hydrolysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR