Heptanoic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Heptylic acid, Enanthic acid

M-Clarity™ quality level = MQ200

8.07582.0100 8.07582.0500 8.07582.0100 8.07582.0500
EM8.07582.0100CS 955.9 CAD
EM8.07582.0100 EM8.07582.0500 CA8.07582.0100 CA8.07582.0500
Heptanoic acid for synthesis, Sigma-Aldrich®
Heptanoic acid
Formula: C₇H₁₄O₂
MDL Number: MFCD00004426
CAS Number: 111-14-8
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Solubility 2.8 g/l (25 °C)
Melting Point -7.5 °C
Molar Mass 130.19 g/mol
Boiling Point 223 °C (1013 hPa)
Vapor Pressure 1.23 hPa (50 °C)
Flash Point 110 °C
Refractive Index 1.4221 (20 °C, 589 nm)
Density 0.92 g/cm3 (20 °C)
Ignition Point 380 °C
Viscosity Kinematic 5.00 mm2/s (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR