9,10-Phenanthrenequinone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.07564.0025 8.07564.0100 8.07564.0025 8.07564.0100
EM8.07564.0025CS 12426.7 CAD
EM8.07564.0025 EM8.07564.0100 CA8.07564.0025 CA8.07564.0100
9,10-Phenanthrenequinone for synthesis, Sigma-Aldrich®
9,10-Phenanthrenequinone
Formula: C₁₄H₈O₂
MDL Number: MFCD00001163
CAS Number: 84-11-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility 7.5 mg/l (25 °C)
Melting Point 203 - 206 °C
Molar Mass 208.21 g/mol
Bulk Density 350 kg/m3
Boiling Point 360 °C
Flash Point 245 °C
Ignition Point 630 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR