1-Propanethiol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: n-Propanethiol, Propyl mercaptan, n-Propyl mercaptan, 1-Mercaptopropane

M-Clarity™ quality level = MQ200

8.07529.0005 8.07529.0250 8.07529.0005 8.07529.0250
EM8.07529.0005CS 4705.24 CAD
EM8.07529.0005 EM8.07529.0250 CA8.07529.0005 CA8.07529.0250
1-Propanethiol for synthesis, Sigma-Aldrich®
1-Propanethiol
Formula: C₃H₈S
MDL Number: MFCD00004900
CAS Number: 107-03-9
UN: 2402
ADR: 3,II

Order Now

Specification Test Results

Solubility 1.9 g/l (25 °C)
Melting Point -113 °C
Molar Mass 76.16 g/mol
Boiling Point 67 - 68 °C (1013 hPa)
Vapor Pressure 165 hPa (20 °C)
Flash Point -34 °C
Refractive Index 1.438 (20 °C, 589 nm)
Density 0.84 g/cm3 (20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR