1-Pentanol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.07500.0100
8.07500.1000
8.07500.2500
8.07500.0100
8.07500.1000
8.07500.2500
EM8.07500.0100CS
951.16
CAD
EM8.07500.0100
EM8.07500.1000
EM8.07500.2500
CA8.07500.0100
CA8.07500.1000
CA8.07500.2500
1-Pentanol for synthesis, Sigma-Aldrich®
1-Pentanol
Formula:
H₃C(CH₂)₃CH₂OH |
MDL Number:
MFCD00002977 CAS Number: 71-41-0 UN: 1105 ADR: 3,III |
|
Specification Test Results
Solubility | 22 g/l (20 °C) |
Melting Point | -79 °C |
Molar Mass | 88.15 g/mol |
Boiling Point | 138 °C (1013 hPa) |
Vapor Pressure | 2.6 hPa (20 °C) |
Flash Point | 49 °C |
Refractive Index | 1.41 (20 °C, 589 nm) |
Explosion Limit | 1.6 %(V) |
Density | 0.815 g/cm3 (20 °C) |
pH | (22 g/l, H2O, 20 °C) neutral |
Ignition Point | 300 °C DIN 51794 |
Evaporation Number | 2.7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...