1,3-Propanediol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
1,3-Propylene glycol, 1,3-Dihydroxy propane
M-Clarity™ quality level = MQ200
8.07481.0005
8.07481.0250
8.07481.1000
8.07481.0005
8.07481.0250
8.07481.1000
EM8.07481.0005CS
3214.75
CAD
EM8.07481.0005
EM8.07481.0250
EM8.07481.1000
CA8.07481.0005
CA8.07481.0250
CA8.07481.1000
1,3-Propanediol for synthesis, Sigma-Aldrich®
1,3-Propanediol
Formula:
HO(CH₂)₃OH MW: 76.1 g/mol |
MDL Number:
MFCD00002949 CAS Number: 504-63-2 |
Specification Test Results
Solubility | 100 g/l (20 °C) |
Melting Point | -32 °C |
Molar Mass | 76.09 g/mol |
Boiling Point | 213 - 215 °C (1013 hPa) |
Vapor Pressure |
|
Refractive Index | 1.4396 (20 °C, 589 nm) |
Explosion Limit | 2.5 %(V) |
Density | 1.05 g/cm3 (20 °C) |
pH | 4.5 - 7.0 (100 g/l, H2O, 20 °C) |
Ignition Point | 405 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...