Nicotinaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Pyridine-3-carboxaldehyde, 3-Pyridinecarboxaldehyde, 3-Pyridinecarbaldehyde

M-Clarity™ quality level = MQ200

8.07468.0050 8.07468.0250 8.07468.0050 8.07468.0250
EM8.07468.0050CS 17604.36 CAD
EM8.07468.0050 EM8.07468.0250 CA8.07468.0050 CA8.07468.0250
Nicotinaldehyde for synthesis, Sigma-Aldrich®
Nicotinaldehyde
Formula: C₆H₅NO
MW: 107.11 g/mol
MDL Number: MFCD00006382
CAS Number: 500-22-1
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 7 °C
Molar Mass 107.11 g/mol
Boiling Point 202 °C
Vapor Pressure 0.3 hPa (20 °C)
Flash Point 84 °C
Refractive Index 1.5493 (20 °C, 589 nm)
Density 1.14 g/cm3 (20 °C)
pH 5.4 (111 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR