Platinum(IV) chloride (57,5% Pt), anhydrous for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Platinum tetrachloride

M-Clarity™ quality level = MQ200

8.07347.0001 8.07347.0005 8.07347.8100 8.07347.0001 8.07347.0005 8.07347.8100
EM8.07347.0001CS 56778.88 CAD
EM8.07347.0001 EM8.07347.0005 EM8.07347.8100 CA8.07347.0001 CA8.07347.0005 CA8.07347.8100
Platinum(IV) chloride (57,5% Pt), anhydrous for synthesis, Sigma-Aldrich®
Platinum (IV) chloride
Formula: Cl₄Pt
MW: 336.89 g/mol
Melting Pt: 370
MDL Number: MFCD00011182
CAS Number: 13454-96-1
UN: 2923
ADR: 8 (6.1) II

Order Now

Specification Test Results

Solubility 587 g/l (25 °C)
Molar Mass 336.89 g/mol
Density 4.3 g/cm3 (20 °C)
pH  - 1 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR