Platinium(IV) oxide hydrate Pt 80% hydrogenation catalyst, for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Platinum(IV) oxide xhydrate, Adam's type catalyst, Platinum dioxide hydrate, Platinum(IV) oxide hydrate, ADAMS' CATALYST
M-Clarity™ quality level = MQ200
8.07346.0001
8.07346.8500
8.07346.0001
8.07346.8500
EM8.07346.0001CS
671206.12
CAD
EM8.07346.0001
EM8.07346.8500
CA8.07346.0001
CA8.07346.8500
Platinium(IV) oxide hydrate Pt 80% hydrogenation catalyst, for synthesis, Sigma-Aldrich®
Platinium (IV) oxide hydrate
Formula:
O₂Pt·H₂O MW: 245.09 Melting Pt: 450 Density: 10.2 |
MDL Number:
MFCD00066964 CAS Number: 52785-06-5 UN: 3085 ADR: 5.1 (8) II Merck Index: 12,07686 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 450 °C |
Molar Mass | 227.08 g/mol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...