p-Phenylenediamine ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
1,4-Diaminobenzene, PPD, 1,4-Phenylenediamine
M-Clarity™ quality level = MQ200
8.07246.0005
8.07246.0250
8.07246.1000
8.07246.9025
8.07246.0005
8.07246.0250
8.07246.1000
8.07246.9025
EM8.07246.0005CS
39487.36
CAD
EM8.07246.0005
EM8.07246.0250
EM8.07246.1000
EM8.07246.9025
CA8.07246.0005
CA8.07246.0250
CA8.07246.1000
CA8.07246.9025
p-Phenylenediamine ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
p-Phenylenediamine
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 267 °C (1013 hPa) Melting Pt: 139…141 °C Density: 1.135 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007901 CAS Number: 106-50-3 UN: 1673 ADR: 6.1,III |
Specification Test Results
Solubility | 47 g/l (25 °C) |
Melting Point | 139 - 141 °C |
Molar Mass | 108.14 g/mol |
Bulk Density | 600 kg/m3 |
Boiling Point | 267 °C (1013 hPa) |
Vapor Pressure | 0.01 hPa (20 °C) |
Flash Point | 156 °C |
Density | 1.135 g/cm3 (20 °C) |
pH | 9 (50 g/l, H2O, 20 °C) |
Ignition Point | 567 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...