1,5-Pentanediol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Pentane-1,5-diol, 1,5-Dihydroxy pentane

M-Clarity™ quality level = MQ200

8.07061.0100 8.07061.0500 8.07061.0100 8.07061.0500
EM8.07061.0100CS 2904.04 CAD
EM8.07061.0100 EM8.07061.0500 CA8.07061.0100 CA8.07061.0500
1,5-Pentanediol for synthesis, Sigma-Aldrich®
1,5-Pentanediol
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 242
Melting Pt: -16
MDL Number: MFCD00002978
CAS Number: 111-29-5
Merck Index: 12,07256

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -16 °C
Molar Mass 104.15 g/mol
Boiling Point 240 - 242 °C (1013 hPa)
Vapor Pressure 0.01 hPa (20 °C)
Flash Point 136 °C
Refractive Index 1.4494 (20 °C, 589 nm)
Explosion Limit 1.3 - 13.1 %(V)
Density 0.99 g/cm3 (20 °C)
pH 7.5 (H2O)
Ignition Point 330 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR