Diethyl oxalate ≥98.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Oxalic acid diethyl ester

M-Clarity™ quality level = MQ200

8.07059.0100 8.07059.0500 8.07059.1000 8.07059.9025 8.07059.0100 8.07059.0500 8.07059.1000 8.07059.9025
EM8.07059.0100CS 2684.42 CAD
EM8.07059.0100 EM8.07059.0500 EM8.07059.1000 EM8.07059.9025 CA8.07059.0100 CA8.07059.0500 CA8.07059.1000 CA8.07059.9025
Diethyl oxalate ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Diethyl oxalate
Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 185.7
Melting Pt: -41
Density: 1.08 g/cm3 (20 °C)
Flash Pt: 76
Storage Temperature: Ambient
MDL Number: MFCD00009119
CAS Number: 95-92-1
UN: 2525
ADR: 6.1 III
Merck Index: 13,03152

Order Now

Specification Test Results

Solubility (20 °C) (slow decomposition)
Melting Point -41 °C
Molar Mass 146.14 g/mol
Boiling Point 185.7 °C (1013 hPa)
Vapor Pressure 0.27 hPa (20 °C)
Flash Point 75 °C
Refractive Index 1.4096 (20 °C, 589 nm)
Explosion Limit 0.42 - 2.67 %(V)
Density 1.08 g/cm3 (20 °C)
Ignition Point 410 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR