Propylene carbonate ≥99% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.07051.0100 8.07051.1000 8.07051.9912 8.07051.0100 8.07051.1000 8.07051.9912
EM8.07051.0100CS 2870.86 CAD
EM8.07051.0100 EM8.07051.1000 EM8.07051.9912 CA8.07051.0100 CA8.07051.1000 CA8.07051.9912
Propylene carbonate ≥99% (by GC) for synthesis, Sigma-Aldrich®
Propylene carbonate
Formula: C₄H₆O₃
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: –49 °C
Density: 1.20 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005385
CAS Number: 108-32-7

Order Now

Specification Test Results

Solubility 240 g/l (20 °C)
Melting Point -49 °C
Molar Mass 102.09 g/mol
Boiling Point 243 °C (1013 hPa)
Vapor Pressure 0.04 hPa (20 °C)
Flash Point 123 °C
Refractive Index 1.4209 (20 °C, 589 nm)
Explosion Limit 1.8 - 14.3 %(V)
Density 1.20 g/cm3 (20 °C)
pH 7.0 (200 g/l, H2O, 20 °C)
Ignition Point 435 °C DIN 51794
Viscosity Kinematic 1.4 mm2/s (43 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR