4-Picoline for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
4-Methylpyridine, γ-Picoline
M-Clarity™ quality level = MQ200
8.07049.0100
8.07049.0500
8.07049.0100
8.07049.0500
EM8.07049.0100CS
1071.24
CAD
EM8.07049.0100
EM8.07049.0500
CA8.07049.0100
CA8.07049.0500
4-Picoline for synthesis, Sigma-Aldrich®
4-Picoline
Formula:
C₆H₇N MW: 93.13 g/mol |
MDL Number:
MFCD00006440 CAS Number: 108-89-4 UN: 2313 ADR: 3,III |
Specification Test Results
Solubility | 1000 g/l (25 °C) |
Melting Point | 3 °C |
Molar Mass | 93.13 g/mol |
Boiling Point | 145 °C (1013 hPa) |
Vapor Pressure | 5.8 hPa (20 °C) |
Flash Point | 40 °C |
Refractive Index | 1.5049 (20 °C, 589 nm) |
Explosion Limit | 1.3 - 8.7 %(V) |
Density | 0.95 g/cm3 (20 °C) |
Ignition Point |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...