(S)-(-)-α-Methylbenzylamine for synthesis, for resolution of racemates, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: (S)-(-)-1-Phenylethylamine, (S)-(-)-α-Phenylethylamine, L(-)-α-Methylbenzylamine

M-Clarity™ quality level = MQ200

8.07047.0025 8.07047.0250 8.07047.0025 8.07047.0250
EM8.07047.0025CS 9340.96 CAD
EM8.07047.0025 EM8.07047.0250 CA8.07047.0025 CA8.07047.0250
(S)-(-)-α-Methylbenzylamine for synthesis, for resolution of racemates, Sigma-Aldrich®
(S)-(-)-α-Methylbenzylamine
Formula: C₈H₁₁N
MW: 121.18 g/mol
MDL Number: MFCD00064406
CAS Number: 2627-86-3
UN: 2922
ADR: 8 (6.1),II

Order Now

Specification Test Results

Solubility 42 g/l (20 °C)
Melting Point -10 °C
Molar Mass 121.18 g/mol
Boiling Point 187 °C (1013 hPa)
Vapor Pressure 0.4 hPa (20 °C)
Flash Point 71 °C
Refractive Index 1.5259 (20 °C, 589 nm)
Density 0.95 g/cm3 (20 °C)
Ignition Point 355 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR