Octylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1-Aminooctane, n-Octylamine, 1-Octylamine

M-Clarity™ quality level = MQ200

8.06917.0251 8.06917.1000 8.06917.0251 8.06917.1000
EM8.06917.0251EA 91.64 CAD
EM8.06917.0251 EM8.06917.1000 CA8.06917.0251 CA8.06917.1000
Octylamine for synthesis, Sigma-Aldrich®
Octylamine
Formula: H₃C(CH₂)₆CH₂NH₂
MW: 129.25 g/mol
Boiling Pt: 180
Melting Pt: -1
Flash Pt: 60
MDL Number: MFCD00008247
CAS Number: 111-86-4
UN: 2734
ADR: 8 (3) I

Order Now

Specification Test Results

Solubility 0.2 g/l (25 °C)
Melting Point -1 °C
Molar Mass 129.24 g/mol
Boiling Point 178 - 179 °C (1013 hPa)
Vapor Pressure 1 hPa (20 °C)
Flash Point 60 °C
Refractive Index 1.429 (20 °C, 589 nm)
Explosion Limit 1.6 - 8.2 %(V)
Density 0.78 g/cm3 (20 °C)
Ignition Point 265 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR